accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COHNOCl

CAS RN 158525-81-6

IUPAC Name: 3-chloro-N-(4-methoxyphenyl)benzamide
Synonyms: 3-chloro-N-(4-methoxyphenyl)benzamide
Molecular Weight (g/mol): 261.71
Molecular Formula: C14H12ClNO2
InChi Key: OASYJKCQPDJQQR-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=O)C2=CC(Cl)=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results