accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15780-02-6

IUPAC Name: tetrapropylazanium perchlorate
Synonyms: tetrapropylammonium perchlorate
Molecular Weight (g/mol): 285.81
Molecular Formula: C12H28ClNO4
InChi Key: LJDKWXHIGFOFDB-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CCC[N+](CCC)(CCC)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results