accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 157701-72-9

NOOFO

CAS RN 157701-72-9

IUPAC Name: 2-fluoro-4-nitrobenzaldehyde
Synonyms: 2-fluoro-4-nitrobenzaldehyde
Molecular Weight (g/mol): 169.11
Molecular Formula: C7H4FNO3
InChi Key: ZFCMKFOFVGHQNE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results