accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 157373-08-5

FClOFF

CAS RN 157373-08-5

IUPAC Name: 2,3,4-trifluorobenzoyl chloride
Synonyms: 2,3,4-trifluorobenzoyl chloride
Molecular Weight (g/mol): 194.54
Molecular Formula: C7H2ClF3O
InChi Key: NXRQXCFBZGIRGN-UHFFFAOYSA-N
SMILES: FC1=CC=C(C(Cl)=O)C(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results