CAS RN 157369-85-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-(-)-Ibuprofen (S)-(+)-Lysinate, TRC
CAS: 157369-85-2 Molecular Formula: C13 H18 O2 . C6 H14 N2 O2 Molecular Weight (g/mol): 352.47 Synonym: L-Lysine, mono[(αR)-α-methyl-4-(2-methylpropyl)benzeneacetate] (9CI),L-Lysine, mono[(R)-α-methyl-4-(2-methylpropyl)benzeneacetate] (ZCI),Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, (R)-, compd. with L-lysine (1:1) (9CI),R-(-)-Ibuprofen L-lysine salt,(R)-Ibuprofen lysinate,(R)-Ibuprofen lysine IUPAC Name: (2S)-2,6-diaminohexanoic acid;(2R)-2-[4-(2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)Cc1ccc(cc1)[C@@H](C)C(=O)O.NCCCC[C@H](N)C(=O)O