accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15733-63-8

Cl

CAS RN 15733-63-8

IUPAC Name: (5-chloropentyl)benzene
Synonyms: (5-chloropentyl)benzene
Molecular Weight (g/mol): 182.69
Molecular Formula: C11H15Cl
InChi Key: UXJLBVYYDZDPBV-UHFFFAOYSA-N
SMILES: ClCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)