accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 157327-51-0

IUPAC Name: 5H,6H,7H-pyrrolo[3,4-d]pyrimidine dihydrochloride
Synonyms: 5H,6H,7H-pyrrolo[3,4-d]pyrimidine dihydrochloride
Molecular Weight (g/mol): 194.06
Molecular Formula: C6H9Cl2N3
InChi Key: VGQSNQPVXPPLEH-UHFFFAOYSA-N
SMILES: Cl.Cl.C1NCC2=NC=NC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results