accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NOOONOOOMn

CAS RN 15710-66-4

IUPAC Name: manganese(2+) hydrate dinitrate
Synonyms: manganese(2+) hydrate dinitrate
Molecular Weight (g/mol): 196.96
Molecular Formula: H2MnN2O7
InChi Key: HBTFASPVVFSRRI-UHFFFAOYSA-N
SMILES: O.[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)