accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 156866-52-3

CH3H3CCH3OOHNO

CAS RN 156866-52-3

IUPAC Name: tert-butyl N-[(4-formylphenyl)methyl]carbamate
Synonyms: tert-butyl N-[(4-formylphenyl)methyl]carbamate
Molecular Weight (g/mol): 235.28
Molecular Formula: C13H17NO3
InChi Key: HRJJBEIFHFVBRT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results