CAS RN 15679-13-7
CAS RN 15679-13-7
IUPAC Name:
4-methyl-2-(propan-2-yl)-1,3-thiazole
Synonyms:
2-isopropyl-4-methyl-1,3-thiazole
Molecular Weight (g/mol):
141.23
Molecular Formula:
C7H11NS
InChi Key:
OFLXNHNYPQPQKW-UHFFFAOYSA-N
SMILES:
CC(C)C1=NC(C)=CS1
1
–
1
of
1
results
1
2-Isopropyl-4-methylthiazole, 98%, Thermo Scientific™
CAS: 15679-13-7 Molecular Formula: C7H11NS Molecular Weight (g/mol): 141.232 MDL Number: MFCD00036561 InChI Key: OFLXNHNYPQPQKW-UHFFFAOYSA-N Synonym: 2-isopropyl-4-methylthiazole,4-methyl-2-1-methylethyl thiazole,2-isopropyl-4-methyl thiazole,thiazole, 2-isopropyl-4-methyl,thiazole, 4-methyl-2-1-methylethyl,unii-nqf7jo3mup,tropical thiazole,2-isopropyl-4-methyl-1,3-thiazole,fema no. 3555,nqf7jo3mup PubChem CID: 61808 IUPAC Name: 4-methyl-2-propan-2-yl-1,3-thiazole SMILES: CC1=CSC(=N1)C(C)C