accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 156727-74-1

H3CNCH3O(R)N(R)CH3NCH2(S)CH3OH3C

CAS RN 156727-74-1

IUPAC Name: 4-[(R)-[(2S,5R)-2,5-dimethyl-4-(prop-2-en-1-yl)piperazin-1-yl](3-methoxyphenyl)methyl]-N,N-diethylbenzamide
Synonyms: 4-[(R)-[(2S,5R)-2,5-dimethyl-4-(prop-2-en-1-yl)piperazin-1-yl](3-methoxyphenyl)methyl]-N,N-diethylbenzamide
Molecular Weight (g/mol): 449.64
Molecular Formula: C28H39N3O2
InChi Key: KQWVAUSXZDRQPZ-UMTXDNHDSA-N
SMILES: CCN(CC)C(=O)C1=CC=C(C=C1)[C@@H](N1C[C@@H](C)N(CC=C)C[C@@H]1C)C1=CC=CC(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results