accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15650-09-6

H2OH2OH2OOClOOOOClOOOSr

CAS RN 15650-09-6

IUPAC Name: strontium(2+) trihydrate diperchlorate
Synonyms: strontium(2+) trihydrate diperchlorate
Molecular Weight (g/mol): 340.56
Molecular Formula: Cl2H6O11Sr
InChi Key: VRRVWIQWZWFYDJ-UHFFFAOYSA-L
SMILES: O.O.O.[Sr++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results