accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15635-87-7

IrOOOOOOH3CCH3CH3H3CH3CH3C

CAS RN 15635-87-7

IUPAC Name: tris(4-oxopent-2-en-2-olate) iridium
Synonyms: iridium acetylacetonate
Molecular Weight (g/mol): 489.55
Molecular Formula: C15H21IrO6
InChi Key: AZFHXIBNMPIGOD-UHFFFAOYSA-K
SMILES: [Ir].CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.CC([O-])=CC(C)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results