accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 156311-83-0

NPNNONNNNFPFFFFF

CAS RN 156311-83-0

IUPAC Name: hexafluoro-λ⁵-phosphanuide; tris(pyrrolidin-1-yl)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})phosphanium
Synonyms: PyAOP reagent
Molecular Weight (g/mol): 521.39
Molecular Formula: C17H27F6N7OP2
InChi Key: CBZAHNDHLWAZQC-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)