accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH2OOH2COOCH2

CAS RN 15625-89-5

IUPAC Name: 2,2-bis[(prop-2-enoyloxy)methyl]butyl prop-2-enoate
Synonyms: trimethylolpropane triacrylate NK ester A tmpt MFM trimethylolpropane triacrylate (product) trimethylol propane triacrylate saret 351 tmpta setalux UV 2241 ogumont T 200 ccris 92
Molecular Weight (g/mol): 296.32
Molecular Formula: C15H20O6
InChi Key: DAKWPKUUDNSNPN-UHFFFAOYSA-N
SMILES: CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results