accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1562-90-9

ClH3CNCH3NHOOOH2NOH

CAS RN 1562-90-9

IUPAC Name: {amino[7-(diethylamino)-3,4-dioxo-4,10-dihydro-3H-phenoxazin-1-yl]methylidene}oxidanium chloride
Synonyms: {amino[7-(diethylamino)-3,4-dioxo-10H-phenoxazin-1-yl]methylidene}oxidanium chloride
Molecular Weight (g/mol): 363.8
Molecular Formula: C17H18ClN3O4
InChi Key: REPMZEQSQQAHJR-UHFFFAOYSA-N
SMILES: [Cl-].CCN(CC)C1=CC=C2NC3=C(OC2=C1)C(=O)C(=O)C=C3C(N)=[OH+]
Color 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)