accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 156185-63-6

CH3H3CCH3OONOH

CAS RN 156185-63-6

IUPAC Name: tert-butyl 4-(3-hydroxypropyl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(3-hydroxypropyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 243.35
Molecular Formula: C13H25NO3
InChi Key: OXPWHPCCUXESFQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CCCO)CC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)