accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 155877-83-1

H3CNNCH3OHOCH3H3CCH3

CAS RN 155877-83-1

IUPAC Name: 4-{2,14,14,17,17-pentamethyl-2,10-diazatetracyclo[9.8.0.0³,⁸.0¹³,¹⁸]nonadeca-1(11),3,5,7,9,12,18-heptaen-9-yl}benzoic acid
Synonyms: 4-{2,14,14,17,17-pentamethyl-2,10-diazatetracyclo[9.8.0.0³,⁸.0¹³,¹⁸]nonadeca-1(11),3,5,7,9,12,18-heptaen-9-yl}benzoic acid
Molecular Weight (g/mol): 438.57
Molecular Formula: C29H30N2O2
InChi Key: YZZAIQOVMHVWBS-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2C(=NC2=C1C=C1C(=C2)C(C)(C)CCC1(C)C)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results