accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NOHOF

CAS RN 1555-11-9

IUPAC Name: 2-fluoro-6H,7H,8H,9H,10H-cyclohepta[b]quinoline-11-carboxylic acid
Synonyms: 2-fluoro-6H,7H,8H,9H,10H-cyclohepta[b]quinoline-11-carboxylic acid
Molecular Weight (g/mol): 259.28
Molecular Formula: C15H14FNO2
InChi Key: HBSUFSHHGHJZSE-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=C(F)C=CC2=NC2=C1CCCCC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results