accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NN

CAS RN 15546-43-7

IUPAC Name: N4,N4,N4',N4'-tetraphenyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4,N4',N4'-tetraphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 488.63
Molecular Formula: C36H28N2
InChi Key: DCZNSJVFOQPSRV-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results