accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15522-69-7

CH3H3CCH3OCH3H3CCH3OHCH3H3CCH3OCH3H3CCH3OHCH3H3CCH3OCH3H3CCH3OHDy(E/Z)(E/Z)(E/Z)

CAS RN 15522-69-7

IUPAC Name: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) dysprosium
Synonyms: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) dysprosium
Molecular Weight (g/mol): 715.34
Molecular Formula: C33H60DyO6
InChi Key: RBHZQBMBFRBKKB-UHFFFAOYSA-N
SMILES: [Dy].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results