accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNO

CAS RN 154927-01-2

IUPAC Name: 1-methyl-5-phenyl-1H-pyrazole-4-carbaldehyde
Synonyms: 1-methyl-5-phenylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: DLMVCAXLQMJYHP-UHFFFAOYSA-N
SMILES: CN1N=CC(C=O)=C1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results