accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

RhIII

CAS RN 15492-38-3

IUPAC Name: rhodium(3+) triiodide
Synonyms: rhodium(3+) triiodide
Molecular Weight (g/mol): 483.62
Molecular Formula: I3Rh
InChi Key: KXAHUXSHRWNTOD-UHFFFAOYSA-K
SMILES: [Rh+3].[I-].[I-].[I-]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results