CAS RN 154765-05-6
CAS RN 154765-05-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MedChemExpress Adrenomedullin (AM) (13-52), human
MedChemExpress Adrenomedullin (AM) (13-52), human is a 40 amino acid peptide, which acts as an endothelium-dependent vasodilator agent.
2
MedChemExpress Adrenomedullin (AM) (13-52), human
MedChemExpress Adrenomedullin (AM) (13-52), human is a 40 amino acid peptide, which acts as an endothelium-dependent vasodilator agent.