accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15474-63-2

IIIDy

CAS RN 15474-63-2

IUPAC Name: dysprosium(3+) triiodide
Synonyms: dysprosium(3+) triiodide
Molecular Weight (g/mol): 543.21
Molecular Formula: DyI3
InChi Key: RZQFCZYXPRKMTP-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Dy+3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results