accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1544-68-9

FNCS

CAS RN 1544-68-9

IUPAC Name: 1-fluoro-4-isothiocyanatobenzene
Synonyms: 1-fluoro-4-isothiocyanatobenzene
Molecular Weight (g/mol): 153.17
Molecular Formula: C7H4FNS
InChi Key: NFIUJHJMCQQYDL-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results