CAS RN 15432-85-6
IUPAC Name:
sodium oxostibinate
Synonyms:
sodium oxostibinate
Molecular Weight (g/mol):
192.75
Molecular Formula:
NaO3Sb
InChi Key:
BMENBXQCFCSBAQ-UHFFFAOYSA-N
SMILES:
[Na+].[O-][Sb](=O)=O
1
–
1
of
1
results
1
Sodium antimonate trihydrate, 98+%, Thermo Scientific™
CAS: 15432-85-6 Molecular Formula: NaSbO3·3H2O MDL Number: MFCD00058775