accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 154039-60-8

H3CNHO(S)HNO(R)H3CH3C(S)OHONHOHH3CCH3CH3

CAS RN 154039-60-8

IUPAC Name: (2S,3R)-N'-[(1S)-2,2-dimethyl-1-(methylcarbamoyl)propyl]-N,2-dihydroxy-3-(2-methylpropyl)butanediamide
Synonyms: marimastat 马立马司他
Molecular Weight (g/mol): 331.41
Molecular Formula: C15H29N3O5
InChi Key: OCSMOTCMPXTDND-OUAUKWLOSA-N
SMILES: CNC(=O)[C@@H](NC(=O)[C@H](CC(C)C)[C@H](O)C(=O)NO)C(C)(C)C
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results