accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 153556-42-4

OOBrF

CAS RN 153556-42-4

IUPAC Name: 4-bromo-3-fluorobenzoate
Synonyms: 4-bromo-3-fluorobenzoate
Molecular Weight (g/mol): 218
Molecular Formula: C7H3BrFO2
InChi Key: RMYOGXPGIDWJLU-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(Br)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results