accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1534-08-3

IUPAC Name: 1-(methylsulfanyl)ethan-1-one
Synonyms: S-methyl thioacetate
Molecular Weight (g/mol): 90.14
Molecular Formula: C3H6OS
InChi Key: OATSQCXMYKYFQO-UHFFFAOYSA-N
SMILES: CSC(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results