accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15310-28-8

IUPAC Name: 1-methyl-4-[(4-methylbenzenesulfonyl)methanesulfonyl]benzene
Synonyms: 1-methyl-4-[(4-methylbenzenesulfonyl)methanesulfonyl]benzene
Molecular Weight (g/mol): 324.41
Molecular Formula: C15H16O4S2
InChi Key: XLOXYWLRAKCDQL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)CS(=O)(=O)C1=CC=C(C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results