accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 153086-78-3

H3CH3CCH3OONHOONH2

CAS RN 153086-78-3

IUPAC Name: tert-butyl N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}carbamate
Synonyms: tert-butyl N-{2-[2-(2-aminoethoxy)ethoxy]ethyl}carbamate
Molecular Weight (g/mol): 248.32
Molecular Formula: C11H24N2O4
InChi Key: OCUICOFGFQENAS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCOCCOCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results