CAS RN 15306-18-0
CAS RN 15306-18-0
IUPAC Name:
2,4,8,10,13,15-hexakis(trifluoromethyl)-1λ³,5,7λ³,11,12λ³,16-hexaoxa-6-aluminaspiro[5.5⁶.5⁶]hexadeca-1,3,7,9,12,14-hexaen-6-yl
Synonyms:
2,4,8,10,13,15-hexakis(trifluoromethyl)-1λ³,5,7λ³,11,12λ³,16-hexaoxa-6-aluminaspiro[5.5⁶.5⁶]hexadeca-1,3,7,9,12,14-hexaen-6-yl
Molecular Weight (g/mol):
651.16
Molecular Formula:
C15H6AlF18O6
InChi Key:
UXBCCTQAQHRGPJ-JVUUZWNBSA-N
SMILES:
—
1
–
1
of
1
results
1
Aluminum hexafluoro-2,4-pentanedionate, 98+%, Thermo Scientific™
CAS: 15306-18-0 Molecular Formula: C15H3AlF18O6 Molecular Weight (g/mol): 648.14 MDL Number: MFCD00050051 InChI Key: SINRSQHVMMWHSI-UHFFFAOYSA-N Synonym: aluminum hexafluoroacetylacetonate,aluminum hexafluoropentanedionate,aluminum hexafluoroacetylacetonate,3z-4-bis 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy alumanyl oxy-1,1,1,5,5,5-hexafluoropent-3-en-2-one IUPAC Name: aluminium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide) SMILES: [Al+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F