accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15304-57-1

IUPAC Name: 9,18,27,36,37,39,40,41-octaaza-38-stannadecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11,13,15,17(41),18,20,22,24,26,28(40),29,31,33,35-nonadecaene
Synonyms: 9,18,27,36,37,39,40,41-octaaza-38-stannadecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11,13,15,17(41),18,20,22,24,26,28(40),29,31,33,35-nonadecaene
Molecular Weight (g/mol): 631.25
Molecular Formula: C32H16N8Sn
InChi Key: LLVONELOQJAYBZ-UHFFFAOYSA-N
SMILES: [Sn]1N2C3=NC4=NC(=NC5=C6C=CC=CC6=C(N=C6N=C(N=C2C2=CC=CC=C32)C2=CC=CC=C62)N15)C1=CC=CC=C41
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results