accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 152864-32-9

OOOOOOOOOOOOEuEu

CAS RN 152864-32-9

IUPAC Name: tris(oxalic acid) dieuropium hydrate
Synonyms: tris(oxalic acid) dieuropium hydrate
Molecular Weight (g/mol): 592.05
Molecular Formula: C6H8Eu2O13
InChi Key: WZYDQGDCICRXQS-UHFFFAOYSA-N
SMILES: O.[Eu].[Eu].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results