accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15244-37-8

OSOOONi

CAS RN 15244-37-8

IUPAC Name: nickel(2+) sulfate
Synonyms: nickel sulfate NiSO4 nickel(II) sulfate (1:1) nickel(II) sulfate
Molecular Weight (g/mol): 154.75
Molecular Formula: NiO4S
InChi Key: LGQLOGILCSXPEA-UHFFFAOYSA-L
SMILES: [Ni++].[O-]S([O-])(=O)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)