accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SNHOOOHOH2CH3CCH3

CAS RN 15214-89-8

IUPAC Name: sodium 2-methyl-2-(prop-2-enamido)propane-1-sulfonate
Synonyms: sodium 2-methyl-2-(prop-2-enamido)propane-1-sulfonate
Molecular Weight (g/mol): 229.23
Molecular Formula: C7H12NNaO4S
InChi Key: FWFUWXVFYKCSQA-UHFFFAOYSA-M
SMILES: [Na+].CC(C)(CS([O-])(=O)=O)NC(=O)C=C
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)