accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOH3CO

CAS RN 15212-03-0

IUPAC Name: 2-methoxyphenyl acetate
Synonyms: guaiacol acetate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: BHJHPYFAYGAPLS-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results