accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1519-46-6

BrPPBrPhPhPhPhPhPh

CAS RN 1519-46-6

IUPAC Name: triphenyl({2-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium hydrobromidyl
Synonyms: triphenyl({2-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium hydrobromidyl
Molecular Weight (g/mol): 708.64
Molecular Formula: C44H38BrP2
InChi Key: ZBGMSQHUFGVZEI-UHFFFAOYSA-N
SMILES: [Br].C(C1=CC=CC=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results