CAS RN 1516-37-6
CAS RN 1516-37-6
IUPAC Name:
(2-methoxyphenyl)thiourea
Synonyms:
2-methoxyphenylthiourea
Molecular Weight (g/mol):
182.24
Molecular Formula:
C8H10N2OS
InChi Key:
HXCHZMHFZXNFIX-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1NC(N)=S