accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1516-17-2

CH3OCH3O(E)(E)

CAS RN 1516-17-2

IUPAC Name: (2E,4E)-hexa-2,4-dien-1-yl acetate
Synonyms: 2,4-hexadienyl acetate
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: PXVKYPFROMBALG-VNKDHWASSA-N
SMILES: C\C=C\C=C\COC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results