accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15155-41-6

IUPAC Name: 4,7-dibromo-2,1,3-benzothiadiazole
Synonyms: 4,7-dibromo-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 293.96
Molecular Formula: C6H2Br2N2S
InChi Key: FEOWHLLJXAECMU-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)C2=NSN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results