accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15119-34-3

OON

CAS RN 15119-34-3

IUPAC Name: 2-oxo-2H-chromene-3-carbonitrile
Synonyms: 2-oxochromene-3-carbonitrile
Molecular Weight (g/mol): 171.16
Molecular Formula: C10H5NO2
InChi Key: QKJALQPLNMEDAV-UHFFFAOYSA-N
SMILES: O=C1OC2=CC=CC=C2C=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results