accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 151096-42-3

FFFClOBrF

CAS RN 151096-42-3

IUPAC Name: 2-bromo-3,4,5,6-tetrafluorobenzoyl chloride
Synonyms: 2-bromo-3,4,5,6-tetrafluorobenzoyl chloride
Molecular Weight (g/mol): 291.42
Molecular Formula: C7BrClF4O
InChi Key: FLVURBOAHUGHCH-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(Cl)=O)C(Br)=C1F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results