accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HONN

CAS RN 151055-79-7

IUPAC Name: {3-[(1H-imidazol-1-yl)methyl]phenyl}methanol
Synonyms: [3-(imidazol-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: TXQZIKDWFOLTSC-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(CN2C=CN=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results