accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15069-92-8

OHNOO

CAS RN 15069-92-8

IUPAC Name: 5-hydroxypyridine-2-carboxylate
Synonyms: 5-hydroxypyridine-2-carboxylate
Molecular Weight (g/mol): 138.1
Molecular Formula: C6H4NO3
InChi Key: CGRRUFNHHQCLDZ-UHFFFAOYSA-M
SMILES: OC1=CC=C(N=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)