accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ON

CAS RN 1503-49-7

IUPAC Name: 4-benzoylbenzonitrile
Synonyms: 4-benzoylbenzonitrile
Molecular Weight (g/mol): 207.23
Molecular Formula: C14H9NO
InChi Key: YSZWJJANSNFQMM-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results