CAS RN 150214-93-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
trans-Haloperidol N-Oxide, TRC
CAS: 150214-93-0 Molecular Formula: C21H23ClFNO3 Molecular Weight (g/mol): 391.86 Synonym: 4-[trans-4-(4-Chlorophenyl)-4-hydroxy-1-oxido-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone,trans-4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone N-Oxide IUPAC Name: (1r,4s)-4-(4-chlorophenyl)-1-(4-(4-fluorophenyl)-4-oxobutyl)-4-hydroxypiperidine 1-oxide SMILES: O=C(CCC[N@+]1([O-])CC[C@](C2=CC=C(Cl)C=C2)(O)CC1)C3=CC=C(F)C=C3