accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOONHH3C

CAS RN 150190-03-7

IUPAC Name: ethyl 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylate
Synonyms: ethyl 2-methyl-6-oxo-1H-pyridine-4-carboxylate
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: WZUAZSMUJJUHMM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=O)NC(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results