accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 150145-89-4

H3C(S)NH3OOOO

CAS RN 150145-89-4

IUPAC Name: 4-[(1S)-1-azaniumyl-1-carboxylatoethyl]benzoate
Synonyms: 4-[(1S)-1-ammonio-1-carboxylatoethyl]benzoate
Molecular Weight (g/mol): 208.19
Molecular Formula: C10H10NO4
InChi Key: DNCAZYRLRMTVSF-JTQLQIEISA-M
SMILES: C[C@@]([NH3+])(C([O-])=O)C1=CC=C(C=C1)C([O-])=O
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results